2-Methyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid

Supplier: Matrix Scientific

020195-500MG
101809-588EA 289.23 USD
101809-588
2-Methyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid
2-Methyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD04092072
CAS Number: 32287-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR