Hexahydro-1H,3H-pyrido[1,2-c][1,3]oxazine

Supplier: Matrix Scientific

020172-1G
101809-528EA 55.38 USD
101809-528
Hexahydro-1H,3H-pyrido[1,2-c][1,3]oxazine
Hexahydro-1H,3H-pyrido[1,2-c][1,3]oxazine

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD06637496
CAS Number: 6049-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR