3-Methyl-4-n-propylpyridine

Supplier: Matrix Scientific

020170-1G
101809-524EA 52.31 USD
101809-524
3-Methyl-4-n-propylpyridine
3-Methyl-4-n-propylpyridine

Hazard: Irritant

Formula: C₉H₁₃N
MDL Number: MFCD06637484
CAS Number: 3478-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR