1,1'-Ethylenebis[4-(3-phenylpropyl)piperidine]

Supplier: Matrix Scientific

020167-1G
101809-518EA 41.54 USD
101809-518
1,1'-Ethylenebis[4-(3-phenylpropyl)piperidine]
1,1'-Ethylenebis[4-(3-phenylpropyl)piperidine]

Hazard: Irritant

Formula: C₃₀H₄₄N₂
MDL Number: MFCD06637428
CAS Number: 865076-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR