2-(Dimethylamino)nicotinaldehyde

Supplier: Matrix Scientific

020166-500MG
101809-516EA 280 USD
101809-516
2-(Dimethylamino)nicotinaldehyde
2-(Dimethylamino)nicotinaldehyde

Hazard: Irritant

Formula: C₈H₁₀N₂O
MDL Number: MFCD07381372
CAS Number: 35567-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR