4-Isobutoxyphenylamine

Supplier: Matrix Scientific

020164-500MG
101809-514EA 280 USD
101809-514
4-Isobutoxyphenylamine
4-Isobutoxyphenylamine

Hazard: Irritant

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 145…146 °C (10 Torr)
MDL Number: MFCD01009861
CAS Number: 5198-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR