4-(4-Allyl-5-hydroxy-3-methylpyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

020162-500MG
101809-510EA 280 USD
101809-510
4-(4-Allyl-5-hydroxy-3-methylpyrazol-1-yl)benzoic acid
4-(4-Allyl-5-hydroxy-3-methylpyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₃
MDL Number: MFCD07793324
CAS Number: 1015844-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR