4-(2,6-Dimethylmorpholino)aniline

Supplier: Matrix Scientific

020144-500MG
101809-476EA 123.08 USD
101809-476
4-(2,6-Dimethylmorpholino)aniline
4-(2,6-Dimethylmorpholino)aniline

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD00051574
CAS Number: 218930-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR