3-Isopropoxyaniline
Supplier: Matrix Scientific
020125-500MG
101809-442EA
153.85
USD
101809-442
3-Isopropoxyaniline
3-Isopropoxyaniline
Hazard: Irritant
Formula:
C₉H₁₃NO MW: 151.21 g/mol |
MDL Number:
MFCD00052700 CAS Number: 41406-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...