2-Cyclopentylethanamine
Supplier: Matrix Scientific
020110-500MG
101809-414EA
136.92
USD
101809-414
2-Cyclopentylethanamine
2-Cyclopentylethanamine
Hazard: Irritant
Formula:
C₇H₁₅N MW: 113.2007 g/mol |
MDL Number:
MFCD00058672 CAS Number: 5763-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...