2-Formyl-3-picoline
Supplier: Matrix Scientific
Synonyms:
3-Methylpyridine-2-carboxaldehyde
, 3-Methylpicolinaldehyde
020072-500MG
101809-348EA
56.92
USD
101809-348
2-Formyl-3-picoline
2-Formyl-3-picoline
Hazard: Irritant
Formula:
C₇H₇NO MW: 121.14 g/mol |
MDL Number:
MFCD02181144 CAS Number: 55589-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...