(4-tert-Butylphenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-(tert-Butyl)phenoxy)acetic acid

019993-500MG
101809-190EA 280 USD
101809-190
(4-tert-Butylphenoxy)acetic acid
(4-tert-Butylphenoxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄(C(CH₃)₃))CH₂COOH
MW: 208.26 g/mol
MDL Number: MFCD00021758
CAS Number: 1798-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR