4-Amino-1-ethyl-1H-pyrazole

Supplier: Matrix Scientific

Synonyms: 1-Ethyl-1H-pyrazol-4-amine

019902-500MG
101809-014EA 72.31 USD
101809-014
4-Amino-1-ethyl-1H-pyrazole
4-Amino-1-ethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₉N₃
MW: 111.1451 g/mol
MDL Number: MFCD05861667
CAS Number: 876343-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR