4-Amino-1-methyl-1H-pyrazole dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1H-pyrazol-4-amine dihydrochloride

019901-1G
101809-012EA 33.35 USD
101809-012
4-Amino-1-methyl-1H-pyrazole dihydrochloride ≥95%
4-Amino-1-methyl-1H-pyrazole dihydrochloride

Hazard: Irritant

Formula: C₄H₉Cl₂N₃
MW: 170.0404 g/mol
MDL Number: MFCD09025437
CAS Number: 1063734-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR