3-[(4-Fluorobenzyl)oxy]-4-methoxybenzaldehyde

Supplier: Matrix Scientific

019877-500MG
101808-966EA 209.3 USD
101808-966
3-[(4-Fluorobenzyl)oxy]-4-methoxybenzaldehyde
3-[(4-Fluorobenzyl)oxy]-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₃FO₃
MDL Number: MFCD02153002
CAS Number: 351066-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR