1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde
Supplier: Matrix Scientific
Synonyms:
1-(Oxan-2-yl)pyrazole-5-carbaldehyde
019788-500MG
101808-794EA
75.38
USD
101808-794
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₉H₁₂N₂O₂ |
MDL Number:
MFCD07186538 CAS Number: 957483-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...