2-Amino-5-methyl-4-phenylthiazole

Supplier: Matrix Scientific

019785-500MG
101808-788EA 11.5 USD
101808-788
2-Amino-5-methyl-4-phenylthiazole
2-Amino-5-methyl-4-phenylthiazole

Hazard: Irritant

Formula: C₁₀H₁₀N₂S
MW: 190.2648 g/mol
MDL Number: MFCD00617257
CAS Number: 30709-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR