2-(2-Methoxyphenoxy)propionic acid

Supplier: Matrix Scientific

019754-500MG
101808-730EA 280 USD
101808-730
2-(2-Methoxyphenoxy)propionic acid
2-(2-Methoxyphenoxy)propionic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₄
MDL Number: MFCD01695530
CAS Number: 7309-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR