3-(1-Piperazinyl)-1,2-benzisothiazole

Supplier: Matrix Scientific

019751-5G 019751-25G
101808-724EA 66.46 USD InStock
101808-724 101808-726
3-(1-Piperazinyl)-1,2-benzisothiazole
3-(1-Piperazinyl)-1,2-benzisothiazole

CAS: 87691-87-0
MDL Number: MFCD04117970
Molecular Formula: C11H13N3S
Molecular Weight: 219,31
Boiling Point: 320°C
Density: 1,256
Flash point: 147°C
Hazard: Irritant

Formula: C₁₁H₁₃N₃S
Boiling Pt: 320 °C
Melting Pt: 86…91 °C
Density: 1,256
Flash Pt: 147 °C
CAS Number: 87691-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR