2-Pyrrolidin-2-yl-1H-benzoimidazole

Supplier: Matrix Scientific

019666-500MG
101808-560EA 280 USD
101808-560
2-Pyrrolidin-2-yl-1H-benzoimidazole
2-Pyrrolidin-2-yl-1H-benzoimidazole

Hazard: Irritant

Formula: C₁₁H₁₃N₃
MDL Number: MFCD01859287
CAS Number: 638141-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR