4-Bromo-1-phenyl-1H-pyrazole

Supplier: Matrix Scientific

019654-500MG
101808-538EA 84.62 USD
101808-538
4-Bromo-1-phenyl-1H-pyrazole
4-Bromo-1-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD00234505
CAS Number: 15115-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR