[1,2,4]Triazol-1-ylacetic acid

Supplier: Matrix Scientific

Synonyms: 2-(1H-1,2,4-Triazol-1-yl)acetic acid

019644-500MG
101808-520EA 126.15 USD
101808-520
[1,2,4]Triazol-1-ylacetic acid
[1,2,4]Triazol-1-ylacetic acid

Hazard: Irritant

Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
MDL Number: MFCD00036051
CAS Number: 28711-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR