5-Phenoxymethyl-4-phenyl-4H-[1,2,4]triazole-3-thiol

Supplier: Matrix Scientific

019631-500MG
101808-494EA 484.62 USD
101808-494
5-Phenoxymethyl-4-phenyl-4H-[1,2,4]triazole-3-thiol
5-Phenoxymethyl-4-phenyl-4H-[1,2,4]triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₃N₃OS
MDL Number: MFCD00619001
CAS Number: 21358-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR