4-Amino-1-isopropylpiperidine
Supplier: Matrix Scientific
019601-500MG
101808-434EA
280
USD
101808-434
4-Amino-1-isopropylpiperidine
4-Amino-1-isopropylpiperidine
Hazard: Irritant
Formula:
C₈H₁₈N₂ MW: 142.2419 g/mol |
MDL Number:
MFCD03411606 CAS Number: 127285-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...