3-(2,6-Dimethylmorpholin-4-yl)propionic acid

Supplier: Matrix Scientific

019573-500MG
101808-382EA 484.62 USD
101808-382
3-(2,6-Dimethylmorpholin-4-yl)propionic acid
3-(2,6-Dimethylmorpholin-4-yl)propionic acid

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD06753041
CAS Number: 883550-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR