C-(4,5,6,7-Tetrahydro-1H-indazol-3-yl)methylamine

Supplier: Matrix Scientific

019560-500MG
101808-356EA 280 USD
101808-356
C-(4,5,6,7-Tetrahydro-1H-indazol-3-yl)methylamine
C-(4,5,6,7-Tetrahydro-1H-indazol-3-yl)methylamine

Hazard: Irritant

Formula: C₈H₁₃N₃
MDL Number: MFCD06753030
CAS Number: 883547-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR