4,5,6-Trimethoxy-1-methyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

019438-500MG
101808-108EA 280 USD
101808-108
4,5,6-Trimethoxy-1-methyl-1H-indole-2-carboxylic acid
4,5,6-Trimethoxy-1-methyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₅
MDL Number: MFCD03848073
CAS Number: 481074-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR