4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid

Supplier: Matrix Scientific

019405-500MG
101808-048EA 280 USD
101808-048
4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid
4-(Tetrahydrofuran-2-ylmethoxy)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MDL Number: MFCD03970016
CAS Number: 565194-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR