2-Chloro-1-(2-methylpiperidin-1-yl)ethanone

Supplier: Matrix Scientific

019398-500MG
101808-034EA 484.62 USD
101808-034
2-Chloro-1-(2-methylpiperidin-1-yl)ethanone
2-Chloro-1-(2-methylpiperidin-1-yl)ethanone

Hazard: Irritant

Formula: C₈H₁₄ClNO
MDL Number: MFCD03030353
CAS Number: 4593-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR