2-(4-Formyl-2-methoxyphenoxy)acetamide

Supplier: Matrix Scientific

019377-500MG
101807-996EA 38.46 USD
101807-996
2-(4-Formyl-2-methoxyphenoxy)acetamide
2-(4-Formyl-2-methoxyphenoxy)acetamide

Hazard: Irritant

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD01050452
CAS Number: 186685-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR