2-Propylsulfanylpyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

019367-500MG
101807-980EA 280 USD
101807-980
2-Propylsulfanylpyrimidine-5-carbaldehyde
2-Propylsulfanylpyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₈H₁₀N₂OS
MW: 182.25 g/mol
MDL Number: MFCD07186427
CAS Number: 876890-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR