MATR019356

019356-5G 019356-25G 019356-1G
101807-958EA 18.31 USD
101807-958 101807-956 101807-960
MATR019356
CAS: 40283-41-8
MDL Number: MFCD00859429
Molecular Formula: C4H4N2O2S
Molecular Weight: 144,15
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR