1-(2,4-Dimethoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

019332-500MG
101807-908EA 746.15 USD
101807-908
1-(2,4-Dimethoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(2,4-Dimethoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₅
MDL Number: MFCD03867080
CAS Number: 133749-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR