(5-Methyl[1,3,4]thiadiazol-2-ylsulfanyl)acetic acid

Supplier: Matrix Scientific

019319-500MG
101807-884EA 461.54 USD
101807-884
(5-Methyl[1,3,4]thiadiazol-2-ylsulfanyl)acetic acid
(5-Methyl[1,3,4]thiadiazol-2-ylsulfanyl)acetic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂S₂
MDL Number: MFCD02257969
CAS Number: 50918-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR