4-(2-Methylimidazol-1-ylmethyl)benzoic acid

Supplier: Matrix Scientific

019289-500MG
101807-830EA 280 USD
101807-830
4-(2-Methylimidazol-1-ylmethyl)benzoic acid
4-(2-Methylimidazol-1-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD07186454
CAS Number: 192637-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR