(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

019277-500MG
101807-806EA 280 USD
101807-806
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₉BrN₂O₂
MDL Number: MFCD00297197
CAS Number: 312309-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR