4'-Methoxybiphenyl-3-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 4'-Methoxy-[1,1'-biphenyl]-3-carboxaldehyde

019266-500MG
101807-786EA 95.38 USD
101807-786
4'-Methoxybiphenyl-3-carbaldehyde
4'-Methoxybiphenyl-3-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD03424627
CAS Number: 118350-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR