4-(2-Methylthiazol-4-yl)phenylamine

Supplier: Matrix Scientific

Synonyms: 4-(2-Methyl-1,3-thiazol-4-yl)aniline

019251-500MG
101807-758EA 280 USD
101807-758
4-(2-Methylthiazol-4-yl)phenylamine
4-(2-Methylthiazol-4-yl)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₀N₂S
MW: 190.27 g/mol
MDL Number: MFCD02691928
CAS Number: 25021-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR