4-Pyridin-4-ylbutyric acid

Supplier: Matrix Scientific

Synonyms: 4-(Pyridin-4-yl)butanoic acid

019159-500MG
101807-590EA 400 USD
101807-590
4-Pyridin-4-ylbutyric acid
4-Pyridin-4-ylbutyric acid

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00673154
CAS Number: 102878-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR