1-Methyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: Matrix Scientific

019150-1G 019150-25G
101807-570EA 20 USD
101807-570 102663-614
1-Methyl-1H-pyrazole-4-carboxylic acid ≥97%
1-Methyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.1100 g/mol
Melting Pt: 203…208 °C
MDL Number: MFCD00159641
CAS Number: 5952-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR