1-[2-(4-Fluorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

019104-500MG
101807-482EA 400 USD
101807-482
1-[2-(4-Fluorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid
1-[2-(4-Fluorophenyl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄FNO₃
MDL Number: MFCD07186388
CAS Number: 842971-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR