2,2-Dimethyl-3-(4-methylpiperidin-1-yl)propionaldehyde

Supplier: Matrix Scientific

019041-500MG
101807-356EA 280 USD
101807-356
2,2-Dimethyl-3-(4-methylpiperidin-1-yl)propionaldehyde
2,2-Dimethyl-3-(4-methylpiperidin-1-yl)propionaldehyde

Hazard: Irritant

Formula: C₁₁H₂₁NO
MW: 183.3 g/mol
MDL Number: MFCD05022484
CAS Number: 690632-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR