(2,5-Dimethyl-1H-indol-3-yl)acetic acid

Supplier: Matrix Scientific

019033-500MG
101807-340EA 378.46 USD
101807-340
(2,5-Dimethyl-1H-indol-3-yl)acetic acid
(2,5-Dimethyl-1H-indol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD02664400
CAS Number: 5435-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR