4-Methoxymethylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-(Methoxymethyl)benzoic acid

019002-500MG
101807-280EA 307.69 USD
101807-280
4-Methoxymethylbenzoic acid
4-Methoxymethylbenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD06375934
CAS Number: 67003-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR