4-Piperidin-3-ylmethylmorpholine

Supplier: Matrix Scientific

018973-500MG
101807-224EA 324.62 USD
101807-224
4-Piperidin-3-ylmethylmorpholine
4-Piperidin-3-ylmethylmorpholine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O
MW: 184.28 g/mol
MDL Number: MFCD05022469
CAS Number: 81310-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR