3,5-Dimethyl-1H-indole-2-carboxylic acid

Supplier: Matrix Scientific

018962-500MG
101807-202EA 280 USD
101807-202
3,5-Dimethyl-1H-indole-2-carboxylic acid
3,5-Dimethyl-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD05864521
CAS Number: 16381-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR