C-(1-Cyclopentylpiperidin-4-yl)methylamine

Supplier: Matrix Scientific

018957-500MG
101807-192EA 400 USD
101807-192
C-(1-Cyclopentylpiperidin-4-yl)methylamine
C-(1-Cyclopentylpiperidin-4-yl)methylamine

Hazard: Irritant

Formula: C₁₁H₂₂N₂
MDL Number: MFCD05864491
CAS Number: 132864-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR