2-(4-Fluorophenyl)-2-pyrrolidin-1-ylethylamine

Supplier: Matrix Scientific

018955-500MG
101807-188EA 324.62 USD
101807-188
2-(4-Fluorophenyl)-2-pyrrolidin-1-ylethylamine
2-(4-Fluorophenyl)-2-pyrrolidin-1-ylethylamine

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD07186362
CAS Number: 876710-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR