2'-Methoxybiphenyl-3-carboxylic acid

Supplier: Matrix Scientific

018907-500MG
101807-100EA 280 USD
101807-100
2'-Methoxybiphenyl-3-carboxylic acid
2'-Methoxybiphenyl-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MDL Number: MFCD03424597
CAS Number: 168618-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR