3-(3,5-Dimethylpiperidin-1-yl)propylamine

Supplier: Matrix Scientific

018904-500MG
101807-094EA 400 USD
101807-094
3-(3,5-Dimethylpiperidin-1-yl)propylamine
3-(3,5-Dimethylpiperidin-1-yl)propylamine

Hazard: Irritant

Formula: C₁₀H₂₂N₂
MDL Number: MFCD03030360
CAS Number: 878657-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR